missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-6-methylphenyl isocyanate, 97%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 1.231 |
| Quantity | 5 g |
| Refractive Index | 1.557 |
| UN Number | UN2206 |
| Beilstein | 2690768 |
| Boiling Point | 79°C to 80°C (2 mmHg) |
| Formula Weight | 167.6 |
| Percent Purity | 97% |
| Chemical Name or Material | 2-Chloro-6-methylphenyl isocyanate |
Safety and Handling
| Hazard Category | H315-H319-H330-H334-H335 |
| Hazard Statement | GHS H Statement H330-H334-H315-H319-H335 Fatal if inhaled. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P260-P264b-P271-P280-P284-P285-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S. |
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only