missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-methylphenyl isocyanate, 97%

Catalog No. AAL1164503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1164503 1 g
AAL1164506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1164503 Supplier Thermo Scientific Chemicals Supplier No. L1164503

CAS: 40398-01-4 | C8H6ClNO | 167.592 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 40398-01-4
Molecular Formula C8H6ClNO
Molecular Weight (g/mol) 167.592
MDL Number MFCD00037030
InChI Key FBTQQNYGMICJQZ-UHFFFAOYSA-N
Synonym 2-chloro-6-methylphenyl isocyanate, 2-chloro-6-methylphenylisocyanate, benzene, 1-chloro-2-isocyanato-3-methyl, acmc-1ake5, 2-chloro-6-methylbenzenisocyanate, 2-chloro 6-methyl phenylisocyanate, 2-chloro 6-methyl phenyl isocyanate, 2-chloro-1-isocyanato-6-methyl benzene, 1-chloro-2-isocyanato-3-methyl-benzene, 6-chloro-2-methylphenylisocyanate
PubChem CID 268123
IUPAC Name 1-chloro-2-isocyanato-3-methylbenzene
SMILES CC1=C(C(=CC=C1)Cl)N=C=O

Specifications

Density 1.231
Quantity 1 g
Refractive Index 1.557
UN Number UN2206
Beilstein 2690768
Boiling Point 79°C to 80°C (2 mmHg)
Formula Weight 167.6
Percent Purity 97%
Chemical Name or Material 2-Chloro-6-methylphenyl isocyanate
Hazard Category H315-H319-H330-H334-H335
Hazard Statement GHS H Statement
H330-H334-H315-H319-H335
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P271-P280-P284-P285-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only