missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-methylnicotinic Acid 98.0+%, TCI America™

Catalog No. p-7125155
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17625G 5 g
C176225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C17625G Supplier TCI America Supplier No. C17625G

Chemical Identifiers

CAS 30529-70-5
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00134165
InChI Key ACQXHCHKMFYDPM-UHFFFAOYSA-N
Synonym 2-chloro-6-methylnicotinic acid, 2-chloro-6-methyl-3-pyridinecarboxylic acid, 2-chloro-6-methylnicotinicacid, 6-methyl-2-chloro nicotinic acid, 3-pyridinecarboxylic acid, 2-chloro-6-methyl, pubchem11019, acmc-209hgo, ksc223m1b, 2-chloro-6-methyl nicotinic acid, 2-chloro-6-methyl-nicotinic acid
PubChem CID 121724
IUPAC Name 2-chloro-6-methylpyridine-3-carboxylic acid
SMILES CC1=NC(=C(C=C1)C(=O)O)Cl

Specifications

Melting Point 154°C
Boiling Point 163°C
Quantity 5 g
Formula Weight 171.58
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-6-methylnicotinic Acid