missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-6-méthylnicotinique Acide 98,0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 154°C |
| Boiling Point | 163°C |
| Quantity | 5 g |
| Formula Weight | 171.58 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Poudre cristalline |
| Chemical Name or Material | 2-Chloro-6-methylnicotinic Acid |
Sécurité et manipulation
| TSCA | No |