missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-methylnicotinic acid, 98+%

Catalog No. AC270810050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC270810050 5 g Glass bottle
AC270810250 25 g Glass bottle
AC270811000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC270810050 Supplier Thermo Scientific Chemicals Supplier No. 270810050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 30529-70-5 | C7H6ClNO2 | 171.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 30529-70-5
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00134165
InChI Key ACQXHCHKMFYDPM-UHFFFAOYSA-N
Synonym 2-chloro-6-methylnicotinic acid, 2-chloro-6-methyl-3-pyridinecarboxylic acid, 2-chloro-6-methylnicotinicacid, 6-methyl-2-chloro nicotinic acid, 3-pyridinecarboxylic acid, 2-chloro-6-methyl, pubchem11019, acmc-209hgo, ksc223m1b, 2-chloro-6-methyl nicotinic acid, 2-chloro-6-methyl-nicotinic acid
PubChem CID 121724
IUPAC Name 2-chloro-6-methylpyridine-3-carboxylic acid
SMILES CC1=NC(=C(C=C1)C(=O)O)Cl

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 157.0°C to 161.0°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 171.58
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-6-methylnicotinic acid
Show More Show Less
EINECSNumber 250-229-7

RUO – Research Use Only