missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-6-methylisonicotinic Acid 98.0+%, TCI America™
$68.97 - $274.02
Chemical Identifiers
| CAS | 25462-85-5 |
|---|---|
| Molecular Formula | C7H6ClNO2 |
| Molecular Weight (g/mol) | 171.58 |
| MDL Number | MFCD00052830 |
| InChI Key | ZGZMEKHQIZSZOH-UHFFFAOYSA-N |
| Synonym | 2-chloro-6-methylisonicotinic acid, 2-chloro-6-picoline-4-carboxylic acid, 4-pyridinecarboxylic acid, 2-chloro-6-methyl, 2-methyl-6-chloropyridine-4-carboxylic acid, 2-chloro-6-methyl-pyridine-4-carboxylic acid, 2-chloro-6-methyl-4-pyridinecarboxylic acid, 2-chloro-6-methyl-isonicotinic acid, 2-chloro-6-methyl-4-pyridinecarboxylicacid, 2-chloro-6-methylisonicotinicacid, pubchem17665 |
| PubChem CID | 141209 |
| IUPAC Name | 2-chloro-6-methylpyridine-4-carboxylic acid |
| SMILES | CC1=NC(=CC(=C1)C(=O)O)Cl |
Chemical Identifiers
| 25462-85-5 | |
| 171.58 | |
| ZGZMEKHQIZSZOH-UHFFFAOYSA-N | |
| 141209 | |
| CC1=NC(=CC(=C1)C(=O)O)Cl |
| C7H6ClNO2 | |
| MFCD00052830 | |
| 2-chloro-6-methylisonicotinic acid, 2-chloro-6-picoline-4-carboxylic acid, 4-pyridinecarboxylic acid, 2-chloro-6-methyl, 2-methyl-6-chloropyridine-4-carboxylic acid, 2-chloro-6-methyl-pyridine-4-carboxylic acid, 2-chloro-6-methyl-4-pyridinecarboxylic acid, 2-chloro-6-methyl-isonicotinic acid, 2-chloro-6-methyl-4-pyridinecarboxylicacid, 2-chloro-6-methylisonicotinicacid, pubchem17665 | |
| 2-chloro-6-methylpyridine-4-carboxylic acid |
Specifications
| 25462-85-5 | |
| White-Yellow | |
| MFCD00052830 | |
| 2-chloro-6-methylisonicotinic acid, 2-chloro-6-picoline-4-carboxylic acid, 4-pyridinecarboxylic acid, 2-chloro-6-methyl, 2-methyl-6-chloropyridine-4-carboxylic acid, 2-chloro-6-methyl-pyridine-4-carboxylic acid, 2-chloro-6-methyl-4-pyridinecarboxylic acid, 2-chloro-6-methyl-isonicotinic acid, 2-chloro-6-methyl-4-pyridinecarboxylicacid, 2-chloro-6-methylisonicotinicacid, pubchem17665 | |
| CC1=NC(=CC(=C1)C(=O)O)Cl | |
| 171.58 | |
| 171.58 | |
| Crystalline Powder |
| 204°C | |
| C7H6ClNO2 | |
| 1 g | |
| ZGZMEKHQIZSZOH-UHFFFAOYSA-N | |
| 2-chloro-6-methylpyridine-4-carboxylic acid | |
| 141209 | |
| ≥98.0% (GC) | |
| 2-Chloro-6-methylisonicotinic Acid |
Safety and Handling
TSCA : No