missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-methylbenzonitrile, 98%

Catalog No. AAL0070603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0070603 1 g
AAL0070606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0070603 Supplier Thermo Scientific Chemicals Supplier No. L0070603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6575-09-3 | C8H6ClN | 151.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6575-09-3
MDL Number MFCD00001782

Specifications

Melting Point 79°C to 83°C
Quantity 1 g
Odor Characteristic
Molecular Formula C8H6ClN
UN Number UN3439
Beilstein 2246414
Synonym benzonitrile, 2-chloro-6-methyl, 2-cyano-3-chlorotoluene, 3-chloro-2-toluonitrile, o-tolunitrile, 6-chloro, 6-chloro-o-tolunitrile, unii-41m65jiw54, 2-chloro-6-methyl benzonitrile, 2-chloro-6-methylbenzenecarbonitrile, 2-cyano3-chlorotoluene, pubchem23832
Boiling Point 143°C (38 mmHg)
InChI Key WQWQHJNUHQEGTN-UHFFFAOYSA-N
SMILES CC1=C(C(=CC=C1)Cl)C#N
IUPAC Name 2-chloro-6-methylbenzonitrile
Molecular Weight (g/mol) 151.59
PubChem CID 23053
Formula Weight 151.6
Percent Purity 98%
Chemical Name or Material 2-Chloro-6-methylbenzonitrile
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 229-499-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only