missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-methylbenzenesulfonyl chloride, 98%

Catalog No. p-7044659
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0762903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0762903 Supplier Thermo Scientific Chemicals Supplier No. L0762903

CAS: 25300-37-2 | C7H6Cl2O2S | 225.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25300-37-2
Molecular Formula C7H6Cl2O2S
Molecular Weight (g/mol) 225.08
MDL Number MFCD00051767
InChI Key FJVZVTWCAGCLCS-UHFFFAOYSA-N
Synonym 2-chloro-6-methylbenzene-1-sulfonyl chloride, 2-chloro-6-methylbenzenesulfonylchloride, 2-chloro-6-methyl-benzenesulfonyl chloride, acmc-20a0m7, chloro 2-chloro-6-methylphenyl sulfone, 2-chloro-6-methylphenylsulfonyl chloride, 2-chloro-6-methylbenzene sulfonyl chloride, 2-chloro-6-methylbenzene-sulfonyl chloride, 2-chloro-6-methylbenzenesulphonyl chloride, benzenesulfonyl chloride, 2-chloro-6-methyl
PubChem CID 15725370
SMILES CC1=C(C(Cl)=CC=C1)S(Cl)(=O)=O

Specifications

Boiling Point 132°C to 134°C (0.5 mmHg)
Refractive Index 1.581
Quantity 1 g
UN Number UN3265
Beilstein 2722659
Sensitivity Moisture sensitive
IUPAC Name 2-chloro-6-methylbenzene-1-sulfonyl chloride
Formula Weight 225.09
Percent Purity 98%
Chemical Name or Material 2-Chloro-6-methylbenzenesulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only