missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-methoxybenzothiazole 96.0+%, TCI America™

Catalog No. C14641G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C14641G 1 g
C14645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C14641G Supplier TCI America Supplier No. C14641G

Chemical Identifiers

CAS 2605-14-3
Molecular Formula C8H6ClNOS
Molecular Weight (g/mol) 199.65
MDL Number MFCD00671510
InChI Key FVUFTABOJFRHSU-UHFFFAOYSA-N
Synonym 2-chloro-6-methoxybenzo d thiazole, 2-chloro-6-methoxybenzothiazole, 2-chloro-6-methoxy-benzothiazole, benzothiazole, 2-chloro-6-methoxy, 6-methoxy-2-chlorobenzothiazole, pubchem17253, acmc-1cipm, chloromethoxybenzothiazole, ksc495g2l, 2-chloro-6-methoxy benzothiazole
PubChem CID 273132
IUPAC Name 2-chloro-6-methoxy-1,3-benzothiazole
SMILES COC1=CC=C2N=C(Cl)SC2=C1

Specifications

Melting Point 54°C
Color White-Yellow
Boiling Point 133°C
Quantity 1 g
Formula Weight 199.65
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-6-methoxybenzothiazole