missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-hydroxypyridine, 98%

Catalog No. AAA1316414
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1316414 25 g
AAA1316422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1316414 Supplier Thermo Scientific Chemicals Supplier No. A1316414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 16879-02-0 | C5H4ClNO | 129.543 g/mol

6-chloro-2-hydroxypyridine was used to study the synthesis, electrochemistry, spectroscopic properties and X-ray crystal structure of Os2Cl(chp)4 (chp = 6-chloro-2- hydroxypyridinate) complex. It undergoes melt reaction with Os2(O2CCH3)4CI2 at 145°C to yield Os2CI4(chp) and Os2Cl(chp)4.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-chloro-2-hydroxypyridine was used to study the synthesis, electrochemistry, spectroscopic properties and X-ray crystal structure of Os2Cl(chp)4 (chp = 6-chloro-2- hydroxypyridinate) complex. It undergoes melt reaction with Os2(O2CCH3)4CI2 at 145°C to yield Os2CI4(chp) and Os2Cl(chp)4.

Notes
Store at room temperature. Ensure adequate ventilation. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 16879-02-0
Molecular Formula C5H4ClNO
Molecular Weight (g/mol) 129.543
MDL Number MFCD00006278
InChI Key CLNNBQDAAGDAHI-UHFFFAOYSA-N
Synonym 6-chloropyridin-2-ol, 2-chloro-6-hydroxypyridine, 6-chloro-2-pyridinol, 6-chloro-2-hydroxypyridine, 6-chloropyridin-2 1h-one, 2 1h-pyridinone, 6-chloro, 6-chloro-2 1h-pyridinone, 6-chloro-2-pyridone, 6-chloropyridn-2-ol, 2 1h-pyridone, 6-chloro
PubChem CID 85621
IUPAC Name 6-chloro-1H-pyridin-2-one
SMILES C1=CC(=O)NC(=C1)Cl

Specifications

Melting Point 129°C to 131°C
Flash Point 163°C (325°F)
Quantity 25 g
Beilstein 108665
Formula Weight 129.55
Percent Purity 98%
Chemical Name or Material 2-Chloro-6-hydroxypyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 240-909-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only