missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-fluorobenzothiazole, 98%

Catalog No. AAH3238303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3238303 1 g
AAH3238306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3238303 Supplier Thermo Scientific Chemicals Supplier No. H3238303

CAS: 399-74-6 | C7H3ClFNS | 187.616 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 399-74-6
Molecular Formula C7H3ClFNS
Molecular Weight (g/mol) 187.616
MDL Number MFCD04448822
InChI Key ISIIQFDYFMPPOA-UHFFFAOYSA-N
Synonym 2-chloro-6-fluorobenzo d thiazole, 2-chloro-6-fluorobenzothiazole, 2-chloro-6-fluoro-benzothiazole, benzothiazole, 2-chloro-6-fluoro, pubchem21812, acmc-1aihk, ksc497m8n, 2-chloro-6-fluoro benzothiazole, benzothiazole, 2-chloro-6-fluoro-8ci,9ci
PubChem CID 2049870
IUPAC Name 2-chloro-6-fluoro-1,3-benzothiazole
SMILES C1=CC2=C(C=C1F)SC(=N2)Cl

Specifications

Melting Point 89°C to 93°C
Quantity 1 g
Formula Weight 187.62
Percent Purity 98%
Chemical Name or Material 2-Chloro-6-fluorobenzothiazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only