missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-fluorobenzonitrile 98.0+%, TCI America™

Catalog No. C10025G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C10025G 5 g
C100225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C10025G Supplier TCI America Supplier No. C10025G
Only null left

Chemical Identifiers

CAS 668-45-1
Molecular Formula C7H3ClFN
Molecular Weight (g/mol) 155.556
MDL Number MFCD00001780
InChI Key XPTAYRHLHAFUOS-UHFFFAOYSA-N
Synonym 2-fluoro-6-chlorobenzonitrile, benzonitrile, 2-chloro-6-fluoro, 2-chloro-6-fluoro-benzonitrile, ncr bg ff, pubchem3423, ksc490q6t, acmc-1b036, timtec-bb sbb042501, 2-chloro-6-fluorobenzenecarbonitrile, 6-chloro-2-fluorobenzenecarbonitrile
PubChem CID 69587
IUPAC Name 2-chloro-6-fluorobenzonitrile
SMILES C1=CC(=C(C(=C1)Cl)C#N)F

Specifications

Melting Point 57°C
Color White-Yellow
Quantity 5 g
Physical Form Crystal-Powder at 20°C
UN Number 3439
Formula Weight 155.56
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2-Chloro-6-fluorobenzonitrile