missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-fluoroaniline 98.0+%, TCI America™

Catalog No. p-7125297
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C21261G 1 g
C21265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C21261G Supplier TCI America Supplier No. C21261G
Only null left

Chemical Identifiers

CAS 363-51-9
Molecular Formula C6H5ClFN
Molecular Weight (g/mol) 145.56
MDL Number MFCD00040309
InChI Key ZJLAWMDJTMMTQB-UHFFFAOYSA-N
Synonym benzenamine, 2-chloro-6-fluoro, 2-chloro-6-fluoro-aniline, 2-chloro-6-fluoro-phenylamine, 6-chloro-2-fluorophenylamine, pubchem1528, 2-fluoro-6-chloroaniline, acmc-209im1, 2-chloro-6-fluoro aniline, 2-chloro-6-fluorobenzenamine, 2-chloro-6-fluorophenylamine
PubChem CID 2734205
IUPAC Name 2-chloro-6-fluoroaniline
SMILES NC1=C(F)C=CC=C1Cl

Specifications

Melting Point 32°C
Color White-Yellow
Quantity 1 g
Physical Form Crystal-Lump at 20°C
Boiling Point 91°C
Formula Weight 145.56
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2-Chloro-6-fluoroaniline