missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-fluoroaniline, 98%

Catalog No. AAA1584803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1584803 1 g
AAA1584806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1584803 Supplier Thermo Scientific Chemicals Supplier No. A1584803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 363-51-9 | C6H5ClFN | 145.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 363-51-9
Molecular Formula C6H5ClFN
Molecular Weight (g/mol) 145.56
MDL Number MFCD00040309
InChI Key ZJLAWMDJTMMTQB-UHFFFAOYSA-N
Synonym benzenamine, 2-chloro-6-fluoro, 2-chloro-6-fluoro-aniline, 2-chloro-6-fluoro-phenylamine, 6-chloro-2-fluorophenylamine, pubchem1528, 2-fluoro-6-chloroaniline, acmc-209im1, 2-chloro-6-fluoro aniline, 2-chloro-6-fluorobenzenamine, 2-chloro-6-fluorophenylamine
PubChem CID 2734205
IUPAC Name 2-chloro-6-fluoroaniline
SMILES NC1=C(F)C=CC=C1Cl

Specifications

Density 1.316
Quantity 1 g
UN Number UN2810
Boiling Point 67°C to 69°C (14 mmHg)
Formula Weight 145.56
Percent Purity 98%
Chemical Name or Material 2-Chloro-6-fluoroaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H373-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 206-657-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only