missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-(trifluoromethyl)pyridine 97.0+%, TCI America™

Catalog No. p-7124848
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C119425G 25 g
C1194100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C119425G Supplier TCI America Supplier No. C119425G

Chemical Identifiers

CAS 52334-81-3
Molecular Formula C6H3ClF3N
Molecular Weight (g/mol) 181.54
MDL Number MFCD00042225
InChI Key JFZJMSDDOOAOIV-UHFFFAOYSA-N
Synonym 2-chloro-5-trifluoromethyl pyridine, 2,5-ctf, pyridine, 2-chloro-5-trifluoromethyl, ccris 6430, 5-trifluoromethyl-2-chloropyridine, unii-c9g66d966b, 2-chloro-5-trifluoropyridine, 2-chloro-5 trifluoromethyl pyridine
PubChem CID 92342
IUPAC Name 2-chloro-5-(trifluoromethyl)pyridine
SMILES FC(F)(F)C1=CC=C(Cl)N=C1

Specifications

Color White-Yellow
Boiling Point 147°C
Quantity 25 g
Formula Weight 181.54
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Chloro-5-(trifluoromethyl)pyridine
EINECSNumber (5)-5431
RTECSNumber US7530000
TSCA No