Learn More
2-Chloro-5-(trifluoromethyl)pyrazine, 97%
CAS: 799557-87-2 | C5H2ClF3N2 | 182.53 g/mol
$125.34 - $206.53
Chemical Identifiers
| CAS | 799557-87-2 |
|---|---|
| Molecular Formula | C5H2ClF3N2 |
| Molecular Weight (g/mol) | 182.53 |
| MDL Number | MFCD13189478 |
| InChI Key | AIEGIFIEQXZBCP-UHFFFAOYSA-N |
| Synonym | 2-chloro-5-trifluoromethyl pyrazine, 5-chloro-2-trifluoromethylpyrazine, pyrazine, 2-chloro-5-trifluoromethyl, 2-chloro-5-trifluoromethyl-pyrazine |
| PubChem CID | 18987645 |
| IUPAC Name | 2-chloro-5-(trifluoromethyl)pyrazine |
| SMILES | C1=C(N=CC(=N1)Cl)C(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH33863MD
|
Thermo Scientific Chemicals
H33863MD |
250 mg |
Each for $125.34
|
|
|||||
|
AAH3386303
|
Thermo Scientific Chemicals
H3386303 |
1 g |
Each for $206.53
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 799557-87-2 | |
| 182.53 | |
| AIEGIFIEQXZBCP-UHFFFAOYSA-N | |
| 18987645 | |
| C1=C(N=CC(=N1)Cl)C(F)(F)F |
| C5H2ClF3N2 | |
| MFCD13189478 | |
| 2-chloro-5-trifluoromethyl pyrazine, 5-chloro-2-trifluoromethylpyrazine, pyrazine, 2-chloro-5-trifluoromethyl, 2-chloro-5-trifluoromethyl-pyrazine | |
| 2-chloro-5-(trifluoromethyl)pyrazine |
Specifications
| 799557-87-2 | |
| C5H2ClF3N2 | |
| 250 mg | |
| 2-chloro-5-trifluoromethyl pyrazine, 5-chloro-2-trifluoromethylpyrazine, pyrazine, 2-chloro-5-trifluoromethyl, 2-chloro-5-trifluoromethyl-pyrazine | |
| C1=C(N=CC(=N1)Cl)C(F)(F)F | |
| 182.53 | |
| 182.53 | |
| 2-Chloro-5-(trifluoromethyl)pyrazine |
| 85°C (112 mmHg) | |
| MFCD13189478 | |
| UN3267 | |
| AIEGIFIEQXZBCP-UHFFFAOYSA-N | |
| 2-chloro-5-(trifluoromethyl)pyrazine | |
| 18987645 | |
| 97% |
Safety and Handling
GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H301+H311+H331-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only