missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-pyridineboronic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 444120-91-6
Molecular Formula C5H5BClNO2
Molecular Weight (g/mol) 157.36
InChI Key WPAPNCXMYWRTTL-UHFFFAOYSA-N
Synonym 2-chloropyridine-5-boronic acid, 6-chloropyridin-3-yl boronic acid, 2-chloro-5-pyridineboronic acid, 2-chloro-5-pyridylboronic acid, 6-chloro-3-pyridylboronic acid, 6-chloropyridin-3-yl-3-boronic acid, 6-chloropyridine-3-boronic acid, 2-chloropyridin-5-ylboronic acid, 6-chloro-3-pyridinylboronic acid, 2-chloro-pyridine-5-boronic acid
PubChem CID 3845034
IUPAC Name (6-chloropyridin-3-yl)boronic acid
SMILES B(C1=CN=C(C=C1)Cl)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC397770010
SDS View Documents
Thermo Scientific Chemicals
397770010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC397770050
SDS View Documents
Thermo Scientific Chemicals
397770050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

444120-91-6
157.36
2-chloropyridine-5-boronic acid, 6-chloropyridin-3-yl boronic acid, 2-chloro-5-pyridineboronic acid, 2-chloro-5-pyridylboronic acid, 6-chloro-3-pyridylboronic acid, 6-chloropyridin-3-yl-3-boronic acid, 6-chloropyridine-3-boronic acid, 2-chloropyridin-5-ylboronic acid, 6-chloro-3-pyridinylboronic acid, 2-chloro-pyridine-5-boronic acid
(6-chloropyridin-3-yl)boronic acid
C5H5BClNO2
WPAPNCXMYWRTTL-UHFFFAOYSA-N
3845034
B(C1=CN=C(C=C1)Cl)(O)O

Specifications

444120-91-6
Glass bottle
1g
WPAPNCXMYWRTTL-UHFFFAOYSA-N
(6-chloropyridin-3-yl)boronic acid
3845034
97%
96% min. (HPLC)
C5H5BClNO2
2-chloropyridine-5-boronic acid, 6-chloropyridin-3-yl boronic acid, 2-chloro-5-pyridineboronic acid, 2-chloro-5-pyridylboronic acid, 6-chloro-3-pyridylboronic acid, 6-chloropyridin-3-yl-3-boronic acid, 6-chloropyridine-3-boronic acid, 2-chloropyridin-5-ylboronic acid, 6-chloro-3-pyridinylboronic acid, 2-chloro-pyridine-5-boronic acid
B(C1=CN=C(C=C1)Cl)(O)O
157.36
157.36
2-Chloro-5-pyridineboronic acid
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only