missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-pyridineboronic acid, 97%

Catalog No. AC397770010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC397770010 1 g
AC397770050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC397770010 Supplier Thermo Scientific Chemicals Supplier No. 397770010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 444120-91-6 | C5H5BClNO2 | 157.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 444120-91-6
Molecular Formula C5H5BClNO2
Molecular Weight (g/mol) 157.36
MDL Number MFCD03094998
InChI Key WPAPNCXMYWRTTL-UHFFFAOYSA-N
Synonym 2-chloropyridine-5-boronic acid, 6-chloropyridin-3-yl boronic acid, 2-chloro-5-pyridineboronic acid, 2-chloro-5-pyridylboronic acid, 6-chloro-3-pyridylboronic acid, 6-chloropyridin-3-yl-3-boronic acid, 6-chloropyridine-3-boronic acid, 2-chloropyridin-5-ylboronic acid, 6-chloro-3-pyridinylboronic acid, 2-chloro-pyridine-5-boronic acid
PubChem CID 3845034
IUPAC Name (6-chloropyridin-3-yl)boronic acid
SMILES OB(O)C1=CC=C(Cl)N=C1

Specifications

Melting Point 165°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 157.36
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-pyridineboronic acid
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only