Learn More
2-Chloro-5-picoline, 94%, Thermo Scientific™
Chemical Identifiers
| CAS | 18368-64-4 |
|---|---|
| Molecular Formula | C6H6ClN |
| Molecular Weight (g/mol) | 127.57 |
| MDL Number | MFCD00792460 |
| InChI Key | VXLYOURCUVQYLN-UHFFFAOYSA-N |
| Synonym | 2-chloro-5-picoline, 6-chloro-3-picoline, pyridine, 2-chloro-5-methyl, 2-chloro-5-methyl-pyridine, 2-chloro-5-methyl pyridine, 3-picoline, 6-chloro, pubchem1193, acmc-1bvpj, 5-methyl-2-chloropyridine |
| PubChem CID | 581393 |
| IUPAC Name | 2-chloro-5-methylpyridine |
| SMILES | CC1=CC=C(Cl)N=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC344030050
|
N/A | 5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC344030250
|
N/A | 25g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 18368-64-4 | |
| 127.57 | |
| VXLYOURCUVQYLN-UHFFFAOYSA-N | |
| 581393 | |
| CC1=CC=C(Cl)N=C1 |
| C6H6ClN | |
| MFCD00792460 | |
| 2-chloro-5-picoline, 6-chloro-3-picoline, pyridine, 2-chloro-5-methyl, 2-chloro-5-methyl-pyridine, 2-chloro-5-methyl pyridine, 3-picoline, 6-chloro, pubchem1193, acmc-1bvpj, 5-methyl-2-chloropyridine | |
| 2-chloro-5-methylpyridine |
Specifications
| 18368-64-4 | |
| 100.0 | |
| 90°C | |
| 92% min. (GC) | |
| 1.5260 to 1.53 | |
| 1.169 | |
| VXLYOURCUVQYLN-UHFFFAOYSA-N | |
| 2-chloro-5-methylpyridine | |
| 581393 | |
| 94% |
| 92.0 | |
| 1.1690g/mL | |
| Authentic | |
| C6H6ClN | |
| MFCD00792460 | |
| 2-chloro-5-picoline, 6-chloro-3-picoline, pyridine, 2-chloro-5-methyl, 2-chloro-5-methyl-pyridine, 2-chloro-5-methyl pyridine, 3-picoline, 6-chloro, pubchem1193, acmc-1bvpj, 5-methyl-2-chloropyridine | |
| CC1=CC=C(Cl)N=C1 | |
| 127.57 | |
| 127.57 | |
| 2-Chloro-5-picoline |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful to aquatic life with long lasting effects.
Causes skin irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO – Research Use Only