missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-5-nitropyridine 99.0+%, TCI America™
$31.68 - $91.33
Chemical Identifiers
| CAS | 4548-45-2 |
|---|---|
| Molecular Formula | C5H3ClN2O2 |
| Molecular Weight (g/mol) | 158.54 |
| MDL Number | MFCD00006240 |
| InChI Key | BAZVFQBTJPBRTJ-UHFFFAOYSA-N |
| Synonym | pyridine, 2-chloro-5-nitro, 2-chloro-5-nitro-pyridine, 3-nitro-6-chloropyridine, chloro 2--5-nitropyridine, 2-chloro-5-nitro pyridine, zlchem 162, pubchem1190, chloro-5-nitropyridine, 2chloro-5-nitropyridine, 2-chloro-5-nitropyridin |
| PubChem CID | 78308 |
| IUPAC Name | 2-chloro-5-nitropyridine |
| SMILES | [O-][N+](=O)C1=CC=C(Cl)N=C1 |
Chemical Identifiers
| 4548-45-2 | |
| 158.54 | |
| BAZVFQBTJPBRTJ-UHFFFAOYSA-N | |
| 78308 | |
| [O-][N+](=O)C1=CC=C(Cl)N=C1 |
| C5H3ClN2O2 | |
| MFCD00006240 | |
| pyridine, 2-chloro-5-nitro, 2-chloro-5-nitro-pyridine, 3-nitro-6-chloropyridine, chloro 2--5-nitropyridine, 2-chloro-5-nitro pyridine, zlchem 162, pubchem1190, chloro-5-nitropyridine, 2chloro-5-nitropyridine, 2-chloro-5-nitropyridin | |
| 2-chloro-5-nitropyridine |
Specifications
| 4548-45-2 | |
| Yellow | |
| MFCD00006240 | |
| pyridine, 2-chloro-5-nitro, 2-chloro-5-nitro-pyridine, 3-nitro-6-chloropyridine, chloro 2--5-nitropyridine, 2-chloro-5-nitro pyridine, zlchem 162, pubchem1190, chloro-5-nitropyridine, 2chloro-5-nitropyridine, 2-chloro-5-nitropyridin | |
| [O-][N+](=O)C1=CC=C(Cl)N=C1 | |
| 158.54 | |
| 158.54 | |
| Crystalline Powder |
| 110°C | |
| C5H3ClN2O2 | |
| 5 g | |
| BAZVFQBTJPBRTJ-UHFFFAOYSA-N | |
| 2-chloro-5-nitropyridine | |
| 78308 | |
| ≥99.0% (GC) | |
| 2-Chloro-5-nitropyridine |
Safety and Handling
RTECSNumber : US7175000
TSCA : No