missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-nitropyridine 99.0+%, TCI America™

Catalog No. C00255G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C00255G 5 g
C002525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C00255G Supplier TCI America Supplier No. C00255G
Only null left

Chemical Identifiers

CAS 4548-45-2
Molecular Formula C5H3ClN2O2
Molecular Weight (g/mol) 158.54
MDL Number MFCD00006240
InChI Key BAZVFQBTJPBRTJ-UHFFFAOYSA-N
Synonym pyridine, 2-chloro-5-nitro, 2-chloro-5-nitro-pyridine, 3-nitro-6-chloropyridine, chloro 2--5-nitropyridine, 2-chloro-5-nitro pyridine, zlchem 162, pubchem1190, chloro-5-nitropyridine, 2chloro-5-nitropyridine, 2-chloro-5-nitropyridin
PubChem CID 78308
IUPAC Name 2-chloro-5-nitropyridine
SMILES [O-][N+](=O)C1=CC=C(Cl)N=C1

Specifications

Melting Point 110°C
Color Yellow
Quantity 5 g
Formula Weight 158.54
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-5-nitropyridine
RTECSNumber US7175000
TSCA No