missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-5-nitropyridine, 99%, Thermo Scientific™

Chemical Identifiers

CAS 4548-45-2
Molecular Formula C5H3ClN2O2
Molecular Weight (g/mol) 158.54
MDL Number MFCD00006240
InChI Key BAZVFQBTJPBRTJ-UHFFFAOYSA-N
Synonym pyridine, 2-chloro-5-nitro, 2-chloro-5-nitro-pyridine, 3-nitro-6-chloropyridine, chloro 2--5-nitropyridine, 2-chloro-5-nitro pyridine, zlchem 162, pubchem1190, chloro-5-nitropyridine, 2chloro-5-nitropyridine, 2-chloro-5-nitropyridin
PubChem CID 78308
IUPAC Name 2-chloro-5-nitropyridine
SMILES [O-][N+](=O)C1=CC=C(Cl)N=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC109740250
SDS View Documents
Thermo Scientific Chemicals
109740250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC109741000
SDS View Documents
Thermo Scientific Chemicals
109741000
100g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

4548-45-2
158.54
BAZVFQBTJPBRTJ-UHFFFAOYSA-N
78308
[O-][N+](=O)C1=CC=C(Cl)N=C1
C5H3ClN2O2
MFCD00006240
pyridine, 2-chloro-5-nitro, 2-chloro-5-nitro-pyridine, 3-nitro-6-chloropyridine, chloro 2--5-nitropyridine, 2-chloro-5-nitro pyridine, zlchem 162, pubchem1190, chloro-5-nitropyridine, 2chloro-5-nitropyridine, 2-chloro-5-nitropyridin
2-chloro-5-nitropyridine

Specifications

4548-45-2
100.0
98.5% min. (GC)
C5H3ClN2O2
MFCD00006240
20, I, 82
Solubility in water: insoluble
[O-][N+](=O)C1=CC=C(Cl)N=C1
158.54
158.54
2-Chloro-5-nitropyridine
98.5
Authentic
Glass bottle
Cl(C5H3N)NO2
25g
pyridine, 2-chloro-5-nitro, 2-chloro-5-nitro-pyridine, 3-nitro-6-chloropyridine, chloro 2--5-nitropyridine, 2-chloro-5-nitro pyridine, zlchem 162, pubchem1190, chloro-5-nitropyridine, 2chloro-5-nitropyridine, 2-chloro-5-nitropyridin
BAZVFQBTJPBRTJ-UHFFFAOYSA-N
2-chloro-5-nitropyridine
78308
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and

GHS Signal Word: Warning

EINECSNumber : 224-908-3

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only