Learn More
2-Chloro-5-nitropyridine, 99%
CAS: 4548-45-2 | C5H3ClN2O2 | 158.54 g/mol
$98.33 - $278.92
Chemical Identifiers
| CAS | 4548-45-2 |
|---|---|
| Molecular Formula | C5H3ClN2O2 |
| Molecular Weight (g/mol) | 158.54 |
| MDL Number | MFCD00006240 |
| InChI Key | BAZVFQBTJPBRTJ-UHFFFAOYSA-N |
| Synonym | pyridine, 2-chloro-5-nitro, 2-chloro-5-nitro-pyridine, 3-nitro-6-chloropyridine, chloro 2--5-nitropyridine, 2-chloro-5-nitro pyridine, zlchem 162, pubchem1190, chloro-5-nitropyridine, 2chloro-5-nitropyridine, 2-chloro-5-nitropyridin |
| PubChem CID | 78308 |
| IUPAC Name | 2-chloro-5-nitropyridine |
| SMILES | [O-][N+](=O)C1=CC=C(Cl)N=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1070914
|
Thermo Scientific Chemicals
A1070914 |
25 g |
Each for $98.33
|
|
|||||
|
AAA1070922
|
Thermo Scientific Chemicals
A1070922 |
100 g |
Each for $278.92
|
|
|||||
Description
2-Chloro-5-nitropyridine is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Chloro-5-nitropyridine is used as a pharmaceutical intermediate.
Solubility
Solubility in hot methanol. Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
| 4548-45-2 | |
| 158.54 | |
| BAZVFQBTJPBRTJ-UHFFFAOYSA-N | |
| 78308 | |
| [O-][N+](=O)C1=CC=C(Cl)N=C1 |
| C5H3ClN2O2 | |
| MFCD00006240 | |
| pyridine, 2-chloro-5-nitro, 2-chloro-5-nitro-pyridine, 3-nitro-6-chloropyridine, chloro 2--5-nitropyridine, 2-chloro-5-nitro pyridine, zlchem 162, pubchem1190, chloro-5-nitropyridine, 2chloro-5-nitropyridine, 2-chloro-5-nitropyridin | |
| 2-chloro-5-nitropyridine |
Specifications
| 4548-45-2 | |
| 158°C (316°F) | |
| MFCD00006240 | |
| 120453 | |
| BAZVFQBTJPBRTJ-UHFFFAOYSA-N | |
| 2-chloro-5-nitropyridine | |
| 78308 | |
| 99% |
| 104°C to 109°C | |
| C5H3ClN2O2 | |
| 25 g | |
| pyridine, 2-chloro-5-nitro, 2-chloro-5-nitro-pyridine, 3-nitro-6-chloropyridine, chloro 2--5-nitropyridine, 2-chloro-5-nitro pyridine, zlchem 162, pubchem1190, chloro-5-nitropyridine, 2chloro-5-nitropyridine, 2-chloro-5-nitropyridin | |
| [O-][N+](=O)C1=CC=C(Cl)N=C1 | |
| 158.54 | |
| 158.54 | |
| 2-Chloro-5-nitropyridine |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 224-908-3
RTECSNumber : US7175000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only