missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-nitropyridine, 99%

Catalog No. AAA1070922
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1070922 100 g
AAA1070914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1070922 Supplier Thermo Scientific Chemicals Supplier No. A1070922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4548-45-2 | C5H3ClN2O2 | 158.54 g/mol

2-Chloro-5-nitropyridine is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-5-nitropyridine is used as a pharmaceutical intermediate.

Solubility
Solubility in hot methanol. Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 4548-45-2
Molecular Formula C5H3ClN2O2
Molecular Weight (g/mol) 158.54
MDL Number MFCD00006240
InChI Key BAZVFQBTJPBRTJ-UHFFFAOYSA-N
Synonym pyridine, 2-chloro-5-nitro, 2-chloro-5-nitro-pyridine, 3-nitro-6-chloropyridine, chloro 2--5-nitropyridine, 2-chloro-5-nitro pyridine, zlchem 162, pubchem1190, chloro-5-nitropyridine, 2chloro-5-nitropyridine, 2-chloro-5-nitropyridin
PubChem CID 78308
IUPAC Name 2-chloro-5-nitropyridine
SMILES [O-][N+](=O)C1=CC=C(Cl)N=C1

Specifications

Melting Point 104°C to 109°C
Flash Point 158°C (316°F)
Quantity 100 g
Beilstein 120453
Formula Weight 158.54
Percent Purity 99%
Chemical Name or Material 2-Chloro-5-nitropyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 224-908-3
RTECSNumber US7175000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only