missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-nitroimidazole, 95%

Catalog No. AAH6114106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6114106 5 g
AAH6114103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6114106 Supplier Thermo Scientific Chemicals Supplier No. H6114106

CAS: 57531-37-0 | C3H2ClN3O2 | 147.518 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 57531-37-0
Molecular Formula C3H2ClN3O2
Molecular Weight (g/mol) 147.518
MDL Number MFCD08705680
InChI Key BOJZBRDIZUHTCE-UHFFFAOYSA-N
Synonym 2-chloro-4-nitroimidazole, 2-chloro-4-nitro-1h-imidazole, 2-chloro-5-nitroimidazole, 1h-imidazole, 2-chloro-4-nitro, 2-chloro-4-nitro-3h-imidazole, nitrochloroimidazole, acmc-1ayku, 2-chloro-4-nitrroimidazole, ksc490o8f
PubChem CID 42409
IUPAC Name 2-chloro-5-nitro-1H-imidazole
SMILES C1=C(NC(=N1)Cl)[N+](=O)[O-]

Specifications

Quantity 5 g
Formula Weight 147.52
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Chloro-5-nitroimidazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 611-554-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only