missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-nitroaniline 98.0+%, TCI America™

Catalog No. p-7124401
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C040325G 25 g
C0403500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C040325G Supplier TCI America Supplier No. C040325G
Only null left

Chemical Identifiers

CAS 6283-25-6
Molecular Formula C6H5ClN2O2
Molecular Weight (g/mol) 172.568
MDL Number MFCD00007668
InChI Key KWIXNFOTNVKIGM-UHFFFAOYSA-N
Synonym benzenamine, 2-chloro-5-nitro, 6-chloro-3-nitroaniline, 2-chloro-5-nitro aniline, aniline, 2-chloro-5-nitro, 2-chloro-5-nitrobenzenamine, aniline, 6-chloro-3-nitro, 2-chloro-5-nitrophenylamine, 2-chloro-5-nitro-benzamine, pubchem21904, 3-nitro-6-chloroaniline
PubChem CID 22691
IUPAC Name 2-chloro-5-nitroaniline
SMILES C1=CC(=C(C=C1[N+](=O)[O-])N)Cl

Specifications

Melting Point 120°C
Quantity 25 g
UN Number 2237
Formula Weight 172.57
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-5-nitroaniline
EINECSNumber (3)-0407
RTECSNumber BX1500000
TSCA Yes