missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-n-pentylpyrimidine, 98%

Catalog No. AAH2780803
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2780803 1 g
AAH2780809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2780803 Supplier Thermo Scientific Chemicals Supplier No. H2780803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 154466-62-3 | C9H13ClN2 | 184.667 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 154466-62-3
Molecular Formula C9H13ClN2
Molecular Weight (g/mol) 184.667
MDL Number MFCD04038093
InChI Key WMARUIMXVALOMF-UHFFFAOYSA-N
Synonym 2-chloro-5-n-pentylpyrimidine, pubchem9038, 5-pentyl-2-chloropyrimidine, 2-chloro-5-pentyl pyrimidine, 2-chloranyl-5-pentyl-pyrimidine
PubChem CID 16756526
IUPAC Name 2-chloro-5-pentylpyrimidine
SMILES CCCCCC1=CN=C(N=C1)Cl

Specifications

Density 1.085
Boiling Point 124°C (3 mmHg)
Flash Point 97°C (207°F)
Refractive Index 1.504
Quantity 1 g
Formula Weight 184.67
Percent Purity 98%
Chemical Name or Material 2-Chloro-5-n-pentylpyrimidine
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only