missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-5-methylthiophene 96.0+%, TCI America™
$63.52 - $309.95
Chemical Identifiers
| CAS | 17249-82-0 |
|---|---|
| Molecular Formula | C5H5ClS |
| Molecular Weight (g/mol) | 132.605 |
| MDL Number | MFCD01632145 |
| InChI Key | JSMMZMYGEVUURX-UHFFFAOYSA-N |
| Synonym | thiophene, 2-chloro-5-methyl, pubchem5197, acmc-209e4p, 2-chloro-5-methyl-thiophene, ksc497i0h, 2-chloranyl-5-methyl-thiophene |
| PubChem CID | 140208 |
| IUPAC Name | 2-chloro-5-methylthiophene |
| SMILES | CC1=CC=C(S1)Cl |
Chemical Identifiers
| 17249-82-0 | |
| 132.605 | |
| JSMMZMYGEVUURX-UHFFFAOYSA-N | |
| 140208 | |
| CC1=CC=C(S1)Cl |
| C5H5ClS | |
| MFCD01632145 | |
| thiophene, 2-chloro-5-methyl, pubchem5197, acmc-209e4p, 2-chloro-5-methyl-thiophene, ksc497i0h, 2-chloranyl-5-methyl-thiophene | |
| 2-chloro-5-methylthiophene |
Specifications
| 17249-82-0 | |
| 155°C | |
| MFCD01632145 | |
| 1993 | |
| JSMMZMYGEVUURX-UHFFFAOYSA-N | |
| 2-chloro-5-methylthiophene | |
| 140208 | |
| ≥96.0% (GC) | |
| 2-Chloro-5-methylthiophene |
| Yellow | |
| C5H5ClS | |
| 5 g | |
| thiophene, 2-chloro-5-methyl, pubchem5197, acmc-209e4p, 2-chloro-5-methyl-thiophene, ksc497i0h, 2-chloranyl-5-methyl-thiophene | |
| CC1=CC=C(S1)Cl | |
| 132.605 | |
| 132.61 | |
| Liquid |
Safety and Handling
TSCA : No