missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-methylthiophene 96.0+%, TCI America™

Catalog No. C23245G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23245G 5 g
C232425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C23245G Supplier TCI America Supplier No. C23245G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17249-82-0
Molecular Formula C5H5ClS
Molecular Weight (g/mol) 132.605
MDL Number MFCD01632145
InChI Key JSMMZMYGEVUURX-UHFFFAOYSA-N
Synonym thiophene, 2-chloro-5-methyl, pubchem5197, acmc-209e4p, 2-chloro-5-methyl-thiophene, ksc497i0h, 2-chloranyl-5-methyl-thiophene
PubChem CID 140208
IUPAC Name 2-chloro-5-methylthiophene
SMILES CC1=CC=C(S1)Cl

Specifications

Color Yellow
Boiling Point 155°C
Quantity 5 g
UN Number 1993
Formula Weight 132.61
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-5-methylthiophene