missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-methylthiophene, 97%

Catalog No. AAB2165706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2165706 5 g
AAB2165714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2165706 Supplier Thermo Scientific Chemicals Supplier No. B2165706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 17249-82-0 | C5H5ClS | 132.605 g/mol

It is used in the synthesis of dithienylcyclopentene optical molecular switches. It is also used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis of dithienylcyclopentene optical molecular switches. It is also used as an active pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix with water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 17249-82-0
Molecular Formula C5H5ClS
Molecular Weight (g/mol) 132.605
MDL Number MFCD01632145
InChI Key JSMMZMYGEVUURX-UHFFFAOYSA-N
Synonym thiophene, 2-chloro-5-methyl, pubchem5197, acmc-209e4p, 2-chloro-5-methyl-thiophene, ksc497i0h, 2-chloranyl-5-methyl-thiophene
PubChem CID 140208
IUPAC Name 2-chloro-5-methylthiophene
SMILES CC1=CC=C(S1)Cl

Specifications

Density 1.21
Boiling Point 155°C to 156°C
Flash Point 47°C (117°F)
Odor Characteristic
Refractive Index 1.5356
Quantity 5 g
UN Number UN1992
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 132.61
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-methylthiophene
Show More Show Less
Hazard Category H226-H302+H312-H331
Hazard Statement GHS H Statement
H331-H226-H302-H312
Toxic if inhaled.
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P311-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only