missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-methylpyrimidine 98.0+%, TCI America™

Catalog No. C29571G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29571G 1 g
C29575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C29571G Supplier TCI America Supplier No. C29571G

Chemical Identifiers

CAS 22536-61-4
Molecular Formula C5H5ClN2
Molecular Weight (g/mol) 128.559
MDL Number MFCD09260903
InChI Key APRMCBSTMFKLEI-UHFFFAOYSA-N
Synonym pyrimidine, 2-chloro-5-methyl, 2-chloro-5-methyl-pyrimidine, pubchem11095, 2-chloro-5-methyl pyrimidine, ksc497i7j
PubChem CID 581719
IUPAC Name 2-chloro-5-methylpyrimidine
SMILES CC1=CN=C(N=C1)Cl

Specifications

Melting Point 94°C
Color Yellow
Quantity 1 g
Formula Weight 128.56
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-5-methylpyrimidine