Learn More
2-Chloro-5-methylpyrimidine, 97%
CAS: 22536-61-4 | C5H5ClN2 | 128.559 g/mol
$181.82 - $706.12
Chemical Identifiers
| CAS | 22536-61-4 |
|---|---|
| Molecular Formula | C5H5ClN2 |
| Molecular Weight (g/mol) | 128.559 |
| MDL Number | MFCD09260903 |
| InChI Key | APRMCBSTMFKLEI-UHFFFAOYSA-N |
| Synonym | pyrimidine, 2-chloro-5-methyl, 2-chloro-5-methyl-pyrimidine, pubchem11095, 2-chloro-5-methyl pyrimidine, ksc497i7j |
| PubChem CID | 581719 |
| IUPAC Name | 2-chloro-5-methylpyrimidine |
| SMILES | CC1=CN=C(N=C1)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6477306
|
Thermo Scientific Chemicals
H6477306 |
5 g |
Each for $181.82
|
|
|||||
|
AAH6477314
|
Thermo Scientific Chemicals
H6477314 |
25 g |
Each for $706.12
|
|
|||||
Description
Used in organic synthesis, catalytic agent, petrochemical additive and synthetic chemistry. Also used as intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed in organic synthesis, catalytic agent, petrochemical additive and synthetic chemistry. Also used as intermediate for pharmaceutical.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Chemical Identifiers
| 22536-61-4 | |
| 128.559 | |
| APRMCBSTMFKLEI-UHFFFAOYSA-N | |
| 581719 | |
| CC1=CN=C(N=C1)Cl |
| C5H5ClN2 | |
| MFCD09260903 | |
| pyrimidine, 2-chloro-5-methyl, 2-chloro-5-methyl-pyrimidine, pubchem11095, 2-chloro-5-methyl pyrimidine, ksc497i7j | |
| 2-chloro-5-methylpyrimidine |
Specifications
| 22536-61-4 | |
| C5H5ClN2 | |
| 5 g | |
| pyrimidine, 2-chloro-5-methyl, 2-chloro-5-methyl-pyrimidine, pubchem11095, 2-chloro-5-methyl pyrimidine, ksc497i7j | |
| APRMCBSTMFKLEI-UHFFFAOYSA-N | |
| 2-chloro-5-methylpyrimidine | |
| 581719 | |
| 97% | |
| 2-Chloro-5-methylpyrimidine |
| 89°C to 92°C | |
| MFCD09260903 | |
| 108732 | |
| Slightly soluble in water. | |
| CC1=CN=C(N=C1)Cl | |
| 128.559 | |
| 128.56 | |
| Solid |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only