missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-methylpyridine 97.0+%, TCI America™

Catalog No. C19995G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C19995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C19995G Supplier TCI America Supplier No. C19995G
Only null left

Chemical Identifiers

CAS 18368-64-4
Molecular Formula C6H6ClN
Molecular Weight (g/mol) 127.57
MDL Number MFCD00792460
InChI Key VXLYOURCUVQYLN-UHFFFAOYSA-N
Synonym 2-chloro-5-picoline, 6-chloro-3-picoline, pyridine, 2-chloro-5-methyl, 2-chloro-5-methyl-pyridine, 2-chloro-5-methyl pyridine, 3-picoline, 6-chloro, pubchem1193, acmc-1bvpj, 5-methyl-2-chloropyridine
PubChem CID 581393
IUPAC Name 2-chloro-5-methylpyridine
SMILES CC1=CC=C(Cl)N=C1

Specifications

Color Yellow
Boiling Point 56°C
Quantity 5 g
Formula Weight 127.57
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-5-methylpyridine
RTECSNumber US6740000
TSCA No