missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-(methylcarbamoyl)benzeneboronic acid, 97%

Catalog No. p-7037056
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH55618MD 250 mg
AAH5561803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH55618MD Supplier Thermo Scientific Chemicals Supplier No. H55618MD

CAS: 2096338-17-7 | C8H9BClNO3 | 213.42 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.

Chemical Identifiers

CAS 2096338-17-7
Molecular Formula C8H9BClNO3
Molecular Weight (g/mol) 213.42
MDL Number MFCD22125158
InChI Key YTEOTIIPHSQWBH-UHFFFAOYSA-N
Synonym 2-chloro-5-methylcarbamoyl benzeneboronic acid, 2-chloro-5-methylcarbamoyl phenylboronic acid
PubChem CID 73996153

Specifications

Quantity 250 mg
Formula Weight 213.43
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-(methylcarbamoyl)benzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only