missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-methyl-3-nitropyridine 98.0+%, TCI America™

Catalog No. C20225G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20225G 5 g
C20221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C20225G Supplier TCI America Supplier No. C20225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 23056-40-8 | C6H5ClN2O2 | 172.568 g/mol

Chemical Identifiers

CAS 23056-40-8
Molecular Formula C6H5ClN2O2
Molecular Weight (g/mol) 172.568
InChI Key LUAJUWOJEFFNFE-UHFFFAOYSA-N
Synonym 2-chloro-3-nitro-5-picoline, 2-chloro-3-nitro-5-methylpyridine, 2-chloro-5-methyl-3-nitro-pyridine, 2-chloro-5-methyl-3-nitro pyridine, pyridine, 2-chloro-5-methyl-3-nitro, pubchem6171, acmc-1ciox, ksc496c3p, 6-chloro-5-nitro-3-picoline
PubChem CID 818300
IUPAC Name 2-chloro-5-methyl-3-nitropyridine
SMILES CC1=CC(=C(N=C1)Cl)[N+](=O)[O-]

Specifications

Melting Point 47°C
Color White-Yellow
Quantity 5 g
Formula Weight 172.57
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-5-methyl-3-nitropyridine