missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-methyl-1,4-phenylenediamine 98.0+%, TCI America™

Supplier:  TCI America D217925G

Catalog No. D217925G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Chloro-5-methyl-1,4-phenylenediamine
5307-03-9
Crystal-Powder at 20°C
MFCD00190684
CPCPKQUNFFHAIZ-UHFFFAOYSA-N
2-chloro-5-methylbenzene-1,4-diamine
79193
≥98.0% (GC,T)
147°C
25 g
C7H9ClN2
2,5-Diamino-4-chlorotoluene
CC1=CC(=C(C=C1N)Cl)N
156.61
156.61

Chemical Identifiers

5307-03-9
156.61
CPCPKQUNFFHAIZ-UHFFFAOYSA-N
79193
CC1=CC(=C(C=C1N)Cl)N
C7H9ClN2
MFCD00190684
2,5-Diamino-4-chlorotoluene
2-chloro-5-methylbenzene-1,4-diamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-3426

TSCA : No

SDS
Documents

Documents

Promotions

Promotions