missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-methoxybenzeneboronic acid, 98%

Catalog No. p-7035640
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5335903 1 g
AAH5335906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5335903 Supplier Thermo Scientific Chemicals Supplier No. H5335903
Only null left

CAS: 89694-46-2 | C7H8BClO3 | 186.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 89694-46-2
Molecular Formula C7H8BClO3
Molecular Weight (g/mol) 186.40
MDL Number MFCD06659858
InChI Key REFXAANPQCJZRY-UHFFFAOYSA-N
Synonym 2-chloro-5-methoxyphenyl boronic acid, 2-chloro-5-methoxyphenylboronicacid, 3-borono-4-chloroanisole, 2-chloro-5-methoxybenzeneboronic acid, 2-chloro-5-methoxy-phenyl boronic acid, boronic acid, 2-chloro-5-methoxyphenyl, pubchem6932, acmc-209r2h, 2-chloro-5-methoxylphenylboronic acid
PubChem CID 17750233
IUPAC Name (2-chloro-5-methoxyphenyl)boronic acid
SMILES COC1=CC=C(Cl)C(=C1)B(O)O

Specifications

Melting Point 106°C to 110°C
Quantity 1 g
Formula Weight 186.4
Percent Purity 98%
Chemical Name or Material 2-Chloro-5-methoxybenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only