missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-iodopyrimidine 98.0+%, TCI America™

Catalog No. C31191G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31191G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C31191G Supplier TCI America Supplier No. C31191G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 32779-38-7
Molecular Formula C4H2ClIN2
Molecular Weight (g/mol) 240.43
MDL Number MFCD06200210
InChI Key WSZRCNZXKKTLQE-UHFFFAOYSA-N
Synonym pyrimidine, 2-chloro-5-iodo, 2-chloro-5-iodo-pyrimidine, pubchem21488, acmc-209hvo, 2-cloro-5-iodopyrimidine, 5-iodo-2-chloropyrimidine, ksc569e7p, pyrimidine,2-chloro-5-iodo
PubChem CID 1714223
IUPAC Name 2-chloro-5-iodopyrimidine
SMILES ClC1=NC=C(I)C=N1

Specifications

Melting Point 135°C
Quantity 1 g
Formula Weight 240.43
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-5-iodopyrimidine