missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-iodopyrimidine, 95%

CAS: 32779-38-7 | C4H2ClIN2 | 240.43 g/mol

Supplier:  Thermo Scientific Chemicals H6342306

 View more versions of this product

Catalog No. AAH6342306

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Chloro-5-iodopyrimidine
C4H2ClIN2
5 g
pyrimidine, 2-chloro-5-iodo, 2-chloro-5-iodo-pyrimidine, pubchem21488, acmc-209hvo, 2-cloro-5-iodopyrimidine, 5-iodo-2-chloropyrimidine, ksc569e7p, pyrimidine,2-chloro-5-iodo
ClC1=NC=C(I)C=N1
240.43
240.43
32779-38-7
MFCD06200210
Light sensitive
WSZRCNZXKKTLQE-UHFFFAOYSA-N
2-chloro-5-iodopyrimidine
1714223
95%

Chemical Identifiers

32779-38-7
240.43
WSZRCNZXKKTLQE-UHFFFAOYSA-N
2-chloro-5-iodopyrimidine
C4H2ClIN2
MFCD06200210
1714223
ClC1=NC=C(I)C=N1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only