missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-iodopyridine 98.0+%, TCI America™

Catalog No. C234125G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C234125G 25 g
C23415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C234125G Supplier TCI America Supplier No. C234125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 69045-79-0
Molecular Formula C5H3ClIN
Molecular Weight (g/mol) 239.44
MDL Number MFCD01863635
InChI Key QWLGCWXSNYKKDO-UHFFFAOYSA-N
Synonym 2-chloro-5-indopyridine, 2-chloro-5-iodo-pyridine, pyridine, 2-chloro-5-iodo, 2-chloro-5-iodo pyridine, zlchem 492, pubchem2078, acmc-1bafa, 2-chloro-5 iodopyridine, 2-chloro-5-iodopyridine, 5-iodo-2-chloro-pyridine
PubChem CID 4397176
IUPAC Name 2-chloro-5-iodopyridine
SMILES C1=CC(=NC=C1I)Cl

Specifications

Melting Point 99°C
Color Yellow
Quantity 25 g
Formula Weight 239.44
Percent Purity ≥98.0% (GC)
Grade Reagent
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-5-iodopyridine