missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-iodopyridine, 98+%, Thermo Scientific™

$318.84 - $1,051.99

Chemical Identifiers

CAS 69045-79-0
Molecular Formula C5H3ClIN
Molecular Weight (g/mol) 239.44
InChI Key QWLGCWXSNYKKDO-UHFFFAOYSA-N
Synonym 2-chloro-5-indopyridine, 2-chloro-5-iodo-pyridine, pyridine, 2-chloro-5-iodo, 2-chloro-5-iodo pyridine, zlchem 492, pubchem2078, acmc-1bafa, 2-chloro-5 iodopyridine, 2-chloro-5-iodopyridine, 5-iodo-2-chloro-pyridine
PubChem CID 4397176
IUPAC Name 2-chloro-5-iodopyridine
SMILES C1=CC(=NC=C1I)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC379070010
View Documents Promotion Details
N/A 1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC379070050
View Documents Promotion Details
N/A 5g Glass bottle
Each for $318.84
Only null left
 
AC379070250
View Documents Promotion Details
N/A 25g Glass bottle
Each for $1,051.99
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

69045-79-0
239.44
2-chloro-5-indopyridine, 2-chloro-5-iodo-pyridine, pyridine, 2-chloro-5-iodo, 2-chloro-5-iodo pyridine, zlchem 492, pubchem2078, acmc-1bafa, 2-chloro-5 iodopyridine, 2-chloro-5-iodopyridine, 5-iodo-2-chloro-pyridine
2-chloro-5-iodopyridine
C5H3ClIN
QWLGCWXSNYKKDO-UHFFFAOYSA-N
4397176
C1=CC(=NC=C1I)Cl

Specifications

69045-79-0
100.0
98% min. (HPLC)
2-chloro-5-indopyridine, 2-chloro-5-iodo-pyridine, pyridine, 2-chloro-5-iodo, 2-chloro-5-iodo pyridine, zlchem 492, pubchem2078, acmc-1bafa, 2-chloro-5 iodopyridine, 2-chloro-5-iodopyridine, 5-iodo-2-chloro-pyridine
QWLGCWXSNYKKDO-UHFFFAOYSA-N
2-chloro-5-iodopyridine
4397176
98+%
98.0
Authentic
C5H3ClIN
Solubility in water: insoluble
C1=CC(=NC=C1I)Cl
239.44
239.44
2-Chloro-5-iodopyridine
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only