missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-iodopyridine, 98%

Catalog No. AAB2527114
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2527114 25 g
AAB2527103 1 g
AAB2527106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2527114 Supplier Thermo Scientific Chemicals Supplier No. B2527114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 69045-79-0 | C5H3ClIN | 239.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 69045-79-0
Molecular Formula C5H3ClIN
Molecular Weight (g/mol) 239.44
MDL Number MFCD01863635
InChI Key QWLGCWXSNYKKDO-UHFFFAOYSA-N
Synonym 2-chloro-5-indopyridine, 2-chloro-5-iodo-pyridine, pyridine, 2-chloro-5-iodo, 2-chloro-5-iodo pyridine, zlchem 492, pubchem2078, acmc-1bafa, 2-chloro-5 iodopyridine, 2-chloro-5-iodopyridine, 5-iodo-2-chloro-pyridine
PubChem CID 4397176
IUPAC Name 2-chloro-5-iodopyridine
SMILES C1=CC(=NC=C1I)Cl

Specifications

Melting Point 95°C to 98°C
Quantity 25 g
Beilstein 108889
Sensitivity Light sensitive
Formula Weight 239.44
Percent Purity 98%
Chemical Name or Material 2-Chloro-5-iodopyridine
Hazard Category H302+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only