missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-hydroxypyridine 97.0+%, TCI America™

Catalog No. C22715G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C22715G Supplier TCI America Supplier No. C22715G
Only null left

Chemical Identifiers

CAS 41288-96-4
Molecular Formula C5H4ClNO
Molecular Weight (g/mol) 129.54
MDL Number MFCD00234050
InChI Key KVCOOWROABTXDJ-UHFFFAOYSA-N
Synonym 2-chloro-5-hydroxypyridine, 6-chloro-3-pyridinol, 3-pyridinol, 6-chloro, 2-chloro-5-hydroxy pyridine, 2-chloro-5-pyridinol, 2-chloropyridin-5-ol, pubchem2410, zlchem 1336, 6-chloropyridine-3-ol, 6-chloro-pyridin-3-ol
PubChem CID 819821
IUPAC Name 6-chloropyridin-3-ol
SMILES OC1=CC=C(Cl)N=C1

Specifications

Melting Point 159°C
Quantity 5 g
Formula Weight 129.54
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-5-hydroxypyridine