missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-fluoropyridine 98.0+%, TCI America™

Catalog No. C22701G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22701G 1 g
C22705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C22701G Supplier TCI America Supplier No. C22701G

Chemical Identifiers

CAS 31301-51-6
Molecular Formula C5H3ClFN
Molecular Weight (g/mol) 131.53
MDL Number MFCD03095332
InChI Key QOGXQLSFJCIDNY-UHFFFAOYSA-N
Synonym 2-chloro-5-fluoro-pyridine, 2-chloro-5-fluoro pyridine, pyridine, 2-chloro-5-fluoro, zlchem 338, pubchem1097, 2-chlor-5-fluorpyridin, acmc-1aevz, 2-chloro-5-fluropyridine, ksc223e3t, tpc-py082
PubChem CID 2782801
IUPAC Name 2-chloro-5-fluoropyridine
SMILES FC1=CN=C(Cl)C=C1

Specifications

Color Yellow
Boiling Point 74°C
Quantity 1 g
UN Number 1993
Formula Weight 131.53
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-5-fluoropyridine