missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-fluoroaniline 98.0+%, TCI America™

Catalog No. C16935G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C16935G Supplier TCI America Supplier No. C16935G

Chemical Identifiers

CAS 452-83-5
Molecular Formula C6H5ClFN
Molecular Weight (g/mol) 145.561
MDL Number MFCD00084852
InChI Key VWUFOZAFKYOZJB-UHFFFAOYSA-N
Synonym 5-fluoro-2-chloroaniline, 2-chloro-5-fluorobenzenamine, 2-chloro-5-fluoro-phenylamine, 2-chloranyl-5-fluoranyl-aniline, pubchem1517, acmc-1bn9k, 2-chloro-5-fluoro-aniline, 2-chloro-5-fluorophenylamine, ksc235o7b, 2-chloro-5-fluorobenzeneamine
PubChem CID 2736509
IUPAC Name 2-chloro-5-fluoroaniline
SMILES C1=CC(=C(C=C1F)N)Cl

Specifications

Color Yellow
Quantity 5 g
Physical Form Clear Liquid at 20°C
Formula Weight 145.56
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2-Chloro-5-fluoroaniline