missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-fluoroaniline, 97%

Catalog No. AAB2561803
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2561803 1 g
AAB2561806 5 g
AAB2561814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2561803 Supplier Thermo Scientific Chemicals Supplier No. B2561803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 452-83-5 | C6H5ClFN | 145.561 g/mol

2-Chloro-5-fluoroaniline is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-5-fluoroaniline is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 452-83-5
Molecular Formula C6H5ClFN
Molecular Weight (g/mol) 145.561
MDL Number MFCD00084852
InChI Key VWUFOZAFKYOZJB-UHFFFAOYSA-N
Synonym 5-fluoro-2-chloroaniline, 2-chloro-5-fluorobenzenamine, 2-chloro-5-fluoro-phenylamine, 2-chloranyl-5-fluoranyl-aniline, pubchem1517, acmc-1bn9k, 2-chloro-5-fluoro-aniline, 2-chloro-5-fluorophenylamine, ksc235o7b, 2-chloro-5-fluorobenzeneamine
PubChem CID 2736509
IUPAC Name 2-chloro-5-fluoroaniline
SMILES C1=CC(=C(C=C1F)N)Cl

Specifications

Melting Point ∼26°C
Quantity 1 g
Odor Characteristic
Refractive Index 1.556
UN Number UN2811
Boiling Point 211°C
Solubility Information Insoluble in water.
Formula Weight 145.56
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-fluoroaniline
Hazard Category H302+H332-H311-H315-H319-H373
Hazard Statement GHS H Statement
H311-H373-H302-H332-H315-H319
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only