missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2'-Chloro-5'-fluoroacetophenone 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 82°C |
| Quantity | 1 g |
| Formula Weight | 172.58 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2′-Chloro-5′-fluoroacetophenone |
Safety and Handling
| TSCA | No |