missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-5-fluoro-3-nitropyridine 98.0+%, TCI America™
$145.40 - $497.70
Chemical Identifiers
| CAS | 136888-21-6 |
|---|---|
| Molecular Formula | C5H2ClFN2O2 |
| Molecular Weight (g/mol) | 176.531 |
| MDL Number | MFCD06659490 |
| InChI Key | SVVZGNAZMPSGMU-UHFFFAOYSA-N |
| PubChem CID | 10773626 |
| IUPAC Name | 2-chloro-5-fluoro-3-nitropyridine |
| SMILES | C1=C(C(=NC=C1F)Cl)[N+](=O)[O-] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
C3115200MG
|
TCI America
C3115200MG |
200 mg |
Each for $145.40
|
|
|||||
|
C31151G
|
TCI America
C31151G |
1 g |
Each for $497.70
|
|
|||||
Chemical Identifiers
| 136888-21-6 | |
| 176.531 | |
| SVVZGNAZMPSGMU-UHFFFAOYSA-N | |
| 2-chloro-5-fluoro-3-nitropyridine |
| C5H2ClFN2O2 | |
| MFCD06659490 | |
| 10773626 | |
| C1=C(C(=NC=C1F)Cl)[N+](=O)[O-] |
Specifications
| 136888-21-6 | |
| Yellow | |
| MFCD06659490 | |
| 2-chloro-3-nitro-5-fluoropyridine, 2-chloro-5-fluoro-3-nitro-pyridine, pyridine, 2-chloro-5-fluoro-3-nitro, pubchem2967, acmc-1cc17, 2-chloro-5-fluoro-3-nitro pyridine, pyridine,2-chloro-5-fluoro-3-nitro | |
| C1=C(C(=NC=C1F)Cl)[N+](=O)[O-] | |
| 176.531 | |
| 176.53 | |
| Crystalline Powder |
| 50°C | |
| C5H2ClFN2O2 | |
| 200 mg | |
| SVVZGNAZMPSGMU-UHFFFAOYSA-N | |
| 2-chloro-5-fluoro-3-nitropyridine | |
| 10773626 | |
| ≥98.0% (GC) | |
| 2-Chloro-5-fluoro-3-nitropyridine |
Safety and Handling
TSCA : No