missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine 98.0+%, TCI America™

Catalog No. p-7125955
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C34361G 1 g
C34365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C34361G Supplier TCI America Supplier No. C34361G
Only null left

Chemical Identifiers

CAS 1003845-08-6
Molecular Formula C10H14BClN2O2
Molecular Weight (g/mol) 240.49
MDL Number MFCD11856048
InChI Key VLAPDEKXZLRRKV-UHFFFAOYSA-N
Synonym 2-chloro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, 2-chloropyrimidine-5-boronic acid pinacol ester, 2-chloro-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, 2-chloropyrimidin-5-ylboronic acid pinacol ester, 2-chloro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyrimidine, 2-chloropyrimidine-5-boronic acid, pinacol ester, 2-chloropyrimidin-5-yl boronic acid pinacol ester, 2-5-chloro-2-pyrimidyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-chloranyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, pyrimidine, 2-chloro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
PubChem CID 51341943
IUPAC Name 2-chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine
SMILES CC1(C)OB(OC1(C)C)C1=CN=C(Cl)N=C1

Specifications

Melting Point 68°C
Color White-Yellow
Quantity 1 g
Formula Weight 240.49
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine