missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4H-1,3,2-benzodioxaphosphorin-4-one 95.0+%, TCI America™

Catalog No. p-7124856
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C12105G 5 g
C121025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C12105G Supplier TCI America Supplier No. C12105G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5381-99-7
Molecular Formula C7H4ClO3P
Molecular Weight (g/mol) 202.53
MDL Number MFCD00013353
InChI Key BVOITXUNGDUXRW-UHFFFAOYNA-N
Synonym 2-chloro-4h-1,3,2-benzodioxaphosphorin-4-one, 2-chloro-4h-benzo d 1,3,2 dioxaphosphinin-4-one, van boom's reagent, 2-chloro-4h-1,2,3-benzodioxaphosphorin-4-one, 4h-1,3,2-benzodioxaphosphorin-4-one, 2-chloro, salicylchlorophosphite, salicyl chlorophosphite, acmc-1an3b, 2-chloro-4h-1,3,2-benzodioxaphosphinin-4-one, salicylchlorophosphite; van boom's reagent
PubChem CID 237010
IUPAC Name 2-chloro-2,4-dihydro-1,3,2-benzodioxaphosphinin-4-one
SMILES ClP1OC(=O)C2=CC=CC=C2O1

Specifications

Melting Point 39°C
Color White-Yellow
Quantity 5 g
UN Number 1759
Formula Weight 202.53
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4H-1,3,2-benzodioxaphosphorin-4-one
TSCA No
Recommended Storage Refrigerator